missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose 97.0+%, TCI America™
$178.95 - $613.27
Chemical Identifiers
| CAS | 22224-41-5 |
|---|---|
| Molecular Formula | C26H22O8 |
| Molecular Weight (g/mol) | 462.454 |
| MDL Number | MFCD00080818 |
| InChI Key | HUHVPBKTTFVAQF-GCUWJAMBSA-N |
| PubChem CID | 68022796 |
| IUPAC Name | [(2R,5R)-3,5-dibenzoyloxy-4-hydroxyoxolan-2-yl]methyl benzoate |
| SMILES | C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)O)OC(=O)C4=CC=CC=C4 |
Chemical Identifiers
| 22224-41-5 | |
| 462.454 | |
| HUHVPBKTTFVAQF-GCUWJAMBSA-N | |
| [(2R,5R)-3,5-dibenzoyloxy-4-hydroxyoxolan-2-yl]methyl benzoate |
| C26H22O8 | |
| MFCD00080818 | |
| 68022796 | |
| C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)O)OC(=O)C4=CC=CC=C4 |
Specifications
| 22224-41-5 | |
| White-Yellow | |
| MFCD00080818 | |
| HUHVPBKTTFVAQF-GCUWJAMBSA-N | |
| [(2R,5R)-3,5-dibenzoyloxy-4-hydroxyoxolan-2-yl]methyl benzoate | |
| 68022796 | |
| ≥97.0% (HPLC) | |
| 1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose |
| 140°C | |
| C26H22O8 | |
| 5 g | |
| C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)O)OC(=O)C4=CC=CC=C4 | |
| 462.454 | |
| 462.45 | |
| Crystalline Powder |
Safety and Handling
TSCA : No