missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose 97.0+%, TCI America™

Catalog No. T26415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26415G 5 g
T264125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26415G Supplier TCI America Supplier No. T26415G

Chemical Identifiers

CAS 22224-41-5
Molecular Formula C26H22O8
Molecular Weight (g/mol) 462.454
MDL Number MFCD00080818
InChI Key HUHVPBKTTFVAQF-GCUWJAMBSA-N
PubChem CID 68022796
IUPAC Name [(2R,5R)-3,5-dibenzoyloxy-4-hydroxyoxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)O)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 5 g
Formula Weight 462.45
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose