missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose, 97%, Thermo Scientific Chemicals

Catalog No. p-7053256
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5946403 1 g
AAH5946406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5946403 Supplier Thermo Scientific Chemicals Supplier No. H5946403
Only null left

It is employed as a starting material for nucleosides and is an intermediate fir cloforabine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a starting material for nucleosides and is an intermediate fir cloforabine.

Solubility
Insoluble in water, easily soluble in chloroform.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.

Chemical Identifiers

CAS 22224-41-5
Molecular Formula C26H22O8
Molecular Weight (g/mol) 462.454
MDL Number MFCD00080818
InChI Key HUHVPBKTTFVAQF-GCUWJAMBSA-N
Synonym α-D-Ribofuranose 1,3,5-tribenzoate
PubChem CID 68022796
IUPAC Name [(2R,5R)-3,5-dibenzoyloxy-4-hydroxyoxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)O)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 125°C to 129°C
Color Brown
Quantity 1 g
Beilstein 98446
Solubility Information Insoluble in water,easily soluble in chloroform.
Formula Weight 462.45
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only