missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri(4-carboxyphenyl)benzene, 97%

Catalog No. AAH6008703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6008703 1 g
AAH6008706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6008703 Supplier Thermo Scientific Chemicals Supplier No. H6008703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50446-44-1 | C27H18O6 | 438.44 g/mol

1,3,5-Tri(4-carboxyphenyl)benzene is used as a building block for metal organic frameworks, which is a 3D-microporous material and finds applications in gas adsorption and separation technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Tri(4-carboxyphenyl)benzene is used as a building block for metal organic frameworks, which is a 3D-microporous material and finds applications in gas adsorption and separation technologies.

Solubility
Soluble in tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents, metals, amines, strong acids, acid chlorides, alcohols, peroxides, permanganates, strong reducing agents, soluble carbonates, phosphates, phosphorus halides and hydroxides.

Chemical Identifiers

CAS 50446-44-1
Molecular Formula C27H18O6
Molecular Weight (g/mol) 438.44
MDL Number MFCD10000888
InChI Key SATWKVZGMWCXOJ-UHFFFAOYSA-N
Synonym 1,3,5-tris 4-carboxyphenyl benzene, 1,3,5-tri 4-carboxyphenyl benzene, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylic acid, 1,1':3',1-terphenyl-4,4-dicarboxylic acid, 5'-4-carboxyphenyl, acmc-209kmn, h3btb, 4,4?,4??,-benzene-1,3,5-triyl-tris benzoic acid, 1,3,5-tris 4-carboxyphenyl benzene, ≤20 wt. % solvent, 3',5'-bis 4-carboxyphenyl-1,1'-biphenyl-4-carboxylic acid, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylicacid
PubChem CID 10694305
IUPAC Name 4-[3,5-bis(4-carboxyphenyl)phenyl]benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)C(O)=O)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 322°C to 327°C
Quantity 1 g
UN Number UN3077
Solubility Information Soluble in tetrahydrofuran.
Formula Weight 438.43
Percent Purity 97%
Chemical Name or Material 1,3,5-Tri(4-carboxyphenyl)benzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only