missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Pentanetricarboxylic Acid 98.0+%, TCI America™

Catalog No. P007525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P007525G 25 g
P00755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P007525G Supplier TCI America Supplier No. P007525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6940-58-5
Molecular Formula C8H12O6
Molecular Weight (g/mol) 204.18
MDL Number MFCD00020546
InChI Key ROTJZTYLACIJIG-UHFFFAOYSA-N
PubChem CID 96220
IUPAC Name pentane-1,3,5-tricarboxylic acid
SMILES OC(=O)CCC(CCC(O)=O)C(O)=O

Specifications

Melting Point 113°C
Color White
Quantity 25 g
Formula Weight 204.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Pentanetricarboxylic Acid