missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3,5-Dichlorophenyl)-1H-pyrrole-2-carbaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 175136-79-5 |
|---|---|
| Molecular Formula | C11H7Cl2NO |
| Molecular Weight (g/mol) | 240.08 |
| MDL Number | MFCD00174281 |
| InChI Key | GNBDQGBCNPLAQK-UHFFFAOYSA-N |
| Synonym | 1-3,5-dichlorophenyl-1h-pyrrole-2-carbaldehyde, 1-3,5-dichlorophenyl pyrrole-2-carbaldehyde, 1-3,5-dichlorophenyl-1h-pyrrole-2-carboxaldehyde, acmc-1c802, n-3,5-dichlorophenyl pyrrole-2-carboxaldehyde, 1-3,5-dichlorophenyl pyrrole-2-carboxaldehyde, 1-3,5-dichloro-phenyl-1h-pyrrole-2-carbald ehyde, 1h-pyrrole-2-carboxaldehyde,1-3,5-dichlorophenyl |
| PubChem CID | 2777016 |
| IUPAC Name | 1-(3,5-dichlorophenyl)pyrrole-2-carbaldehyde |
| SMILES | ClC1=CC(=CC(Cl)=C1)N1C=CC=C1C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
CD08850DA
|
Thermo Scientific Chemicals
CD08850DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CD08850EA
|
Thermo Scientific Chemicals
CD08850EA |
10g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 175136-79-5 | |
| 240.08 | |
| GNBDQGBCNPLAQK-UHFFFAOYSA-N | |
| 2777016 | |
| ClC1=CC(=CC(Cl)=C1)N1C=CC=C1C=O |
| C11H7Cl2NO | |
| MFCD00174281 | |
| 1-3,5-dichlorophenyl-1h-pyrrole-2-carbaldehyde, 1-3,5-dichlorophenyl pyrrole-2-carbaldehyde, 1-3,5-dichlorophenyl-1h-pyrrole-2-carboxaldehyde, acmc-1c802, n-3,5-dichlorophenyl pyrrole-2-carboxaldehyde, 1-3,5-dichlorophenyl pyrrole-2-carboxaldehyde, 1-3,5-dichloro-phenyl-1h-pyrrole-2-carbald ehyde, 1h-pyrrole-2-carboxaldehyde,1-3,5-dichlorophenyl | |
| 1-(3,5-dichlorophenyl)pyrrole-2-carbaldehyde |
Specifications
| 175136-79-5 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD00174281 | |
| 1-3,5-dichlorophenyl-1h-pyrrole-2-carbaldehyde, 1-3,5-dichlorophenyl pyrrole-2-carbaldehyde, 1-3,5-dichlorophenyl-1h-pyrrole-2-carboxaldehyde, acmc-1c802, n-3,5-dichlorophenyl pyrrole-2-carboxaldehyde, 1-3,5-dichlorophenyl pyrrole-2-carboxaldehyde, 1-3,5-dichloro-phenyl-1h-pyrrole-2-carbald ehyde, 1h-pyrrole-2-carboxaldehyde,1-3,5-dichlorophenyl | |
| ClC1=CC(=CC(Cl)=C1)N1C=CC=C1C=O | |
| 240.08 | |
| 240.09 | |
| 1-(3, 5-Dichlorophenyl)-1H-pyrrole-2-carbaldehyde |
| 96.5 | |
| 97% | |
| C11H7Cl2NO | |
| 1g | |
| GNBDQGBCNPLAQK-UHFFFAOYSA-N | |
| 1-(3,5-dichlorophenyl)-1H-pyrrole-2-carbaldehyde | |
| 2777016 | |
| 97% |