missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Cyclohexanetricarbonitrile (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7125411
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C23271G Supplier TCI America Supplier No. C23271G
Only null left

Chemical Identifiers

CAS 183582-92-5
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
MDL Number MFCD08276327
InChI Key VMUOSHREZKXCIV-UHFFFAOYSA-N
Synonym 1,3,5-Tricyanocyclohexane
PubChem CID 11423741
IUPAC Name cyclohexane-1,3,5-tricarbonitrile
SMILES C1C(CC(CC1C#N)C#N)C#N

Specifications

Color White
Boiling Point 200°C
Quantity 1 g
UN Number 3439
Formula Weight 159.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Cyclohexanetricarbonitrile (cis- and trans- mixture)