missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Cycloheptatriene 95.0+%, TCI America™

Catalog No. C057225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C057225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C057225ML Supplier TCI America Supplier No. C057225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 544-25-2
Molecular Formula C7H8
Molecular Weight (g/mol) 92.141
MDL Number MFCD00004146
InChI Key CHVJITGCYZJHLR-UHFFFAOYSA-N
Synonym cycloheptatriene, tropilidene, 1,3,5-cycloheptatriene, tropilidine, tropiliden, cycloheptatrien, tropyliden, zykloheptatrien, 1h-7 annulene, tropilidin
PubChem CID 11000
ChEBI CHEBI:37519
IUPAC Name cyclohepta-1,3,5-triene
SMILES C1C=CC=CC=C1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 mL
UN Number 2603
Formula Weight 92.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Cycloheptatriene
EINECSNumber (9)-0884
RTECSNumber GU3675000
TSCA No
Recommended Storage Refrigerator