missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1,3,5-Benzenetricarbonyl trichloride

Catalog No. 147532500G
Encompass
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
147532500G 500 g
14753225KG 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 147532500G Supplier Sigma Aldrich Supplier No. 147532500G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3,5-Benzenetricarbonyl trichloride

Specifications

CAS 4422-95-1
Melting Point 32°C to 38°C
Density 1.487 g/mL (at 25°C)
Boiling Point 180°C (6 mmHg)
Molecular Formula C9H3Cl3O3
Linear Formula C6H3(COCl)3
MDL Number MFCD00000679
Quantity 500 g
Synonym Benzene-1,3,5-tricarbonyl chloride; Trimesic acid trichloride; Trimesoyl chloride
Molecular Weight (g/mol) 265.48
Percent Purity 98%
Show More Show Less