missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarbonyl chloride, 98+%

Catalog No. AAL0725506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0725506 5 g
AAL0725514 25 g
AAL0725522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0725506 Supplier Thermo Scientific Chemicals Supplier No. L0725506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4422-95-1 | C9H3Cl3O3 | 265.47 g/mol

1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.

Solubility
Soluble in chloroform(soluble10%, clear to very slightly hazy, colorless to faintly yellow) and toluene. Solubility in water: Reacts.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases and oxidizing agents. Store at 4°C.

Chemical Identifiers

CAS 4422-95-1
Molecular Formula C9H3Cl3O3
Molecular Weight (g/mol) 265.47
MDL Number MFCD00000679
InChI Key UWCPYKQBIPYOLX-UHFFFAOYSA-N
Synonym trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
PubChem CID 78138
SMILES ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O

Specifications

Melting Point 34°C to 38°C
Boiling Point 180°C (16 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 2940936
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform(soluble10%,clear to very slightly hazy,colorless to faintly yellow) and toluene. Solubility in water: Reacts.
IUPAC Name benzene-1,3,5-tricarbonyl trichloride
Formula Weight 265.48
Percent Purity ≥98%
Chemical Name or Material 1,3,5-Benzenetricarbonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 224-594-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only