missing translation for 'onlineSavingsMsg'
Learn More

1,3,5,7-Cyclooctatetraene (stabilized with HQ) 98.0+%, TCI America™

Catalog No. C05051ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05051ML 1 mL
C05055ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05051ML Supplier TCI America Supplier No. C05051ML

Chemical Identifiers

CAS 629-20-9
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00004161
InChI Key KDUIUFJBNGTBMD-DLMDZQPMSA-N
Synonym 1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n
PubChem CID 637866
IUPAC Name (1Z,3Z,5Z,7Z)-cycloocta-1,3,5,7-tetraene
SMILES C1=C/C=C\C=C/C=C\1

Specifications

Melting Point -3°C
Color Yellow
Boiling Point 143°C
Quantity 1 mL
UN Number 2358
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5,7-Cyclooctatetraene (stabilized with HQ)
RTECSNumber GY0175600
TSCA Yes
Recommended Storage Refrigerator