missing translation for 'onlineSavingsMsg'
Learn More

1,3,5,7-Cyclooctatetraene, 98%, stabilized

Catalog No. AC160570025
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160570025 2.5 g Glass bottle
AC160570100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160570025 Supplier Thermo Scientific Chemicals Supplier No. 160570025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 629-20-9 | C8H8 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-20-9
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00004161
InChI Key KDUIUFJBNGTBMD-DLMDZQPMSA-N
Synonym 1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n
PubChem CID 637866
IUPAC Name cyclooctatetraene
SMILES C1=C/C=C\C=C/C=C\1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -27°C
Color Brown-Yellow to Yellow
Density 0.9200g/mL
Boiling Point 142°C to 143°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5365 to 1.5385
Quantity 2.5 g
Beilstein 05, I, 228
Specific Gravity 0.92
Merck Index 15, 2731
Solubility Information Solubility in water: immiscible
Stabilizer 0.1% hydroquinone
Formula Weight 104.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3,5,7-Cyclooctatetraene, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cauti
EINECSNumber 211-080-3

RUO – Research Use Only