missing translation for 'onlineSavingsMsg'
Learn More

1,3,5,7-Cyclooctatetraene, 98%, stab. with 0.1% Hydroquinone

Catalog No. AAB2263606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2263606 5 g
AAB2263614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2263606 Supplier Thermo Scientific Chemicals Supplier No. B2263606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 629-20-9 | C8H8 | 104.15 g/mol

1,3,5,7-Cyclooctatetraene is used in the synthesis of highly organic film for silicon surfaces to improve its chemical and physical properties. It is also used in liquid state organic dye lasers, as triplet state quencher to reduce dye blinking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5,7-Cyclooctatetraene is used in the synthesis of highly organic film for silicon surfaces to improve its chemical and physical properties. It is also used in liquid state organic dye lasers, as triplet state quencher to reduce dye blinking.

Solubility
Not miscible or difficult to mix in water.

Notes
Air & light sensitive. Store at --20°C. Store away from oxidizing agent, air, heat, ultra violet radiation, free radical intiators.

Chemical Identifiers

CAS 629-20-9
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00004161
InChI Key KDUIUFJBNGTBMD-DLMDZQPMSA-N
Synonym 1,3,5,7-cyclooctatetraene, 8 annulene, unii-aj19r479cq, cycloocta-1,3,5,7-tetraene, 8-annulene, cyclooctatetraene, 1,5,7-cyclooctatetraene, un2358 flammable liquid, kduiufjbngtbmd-bonzmoemsa-n
PubChem CID 637866
IUPAC Name cyclooctatetraene
SMILES C1=C/C=C\C=C/C=C\1

Specifications

Melting Point -5°C
Density 0.925
Boiling Point 142°C to 143°C
Flash Point 20°C (68°F)
Odor Unpleasant
Refractive Index 1.537
Quantity 5 g
UN Number UN2358
Beilstein 2496800
Sensitivity Air and light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 104.15
Percent Purity 98%
Chemical Name or Material 1,3,5,7-Cyclooctatetraene, stabilized with 0.1% Hydroquinone
Show More Show Less
Hazard Category H225-H304-H315-H319-H335
Hazard Statement GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOOCTATETRAENE
EINECSNumber 211-080-3
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only