missing translation for 'onlineSavingsMsg'
Learn More

1,3,5,5-Tetramethyl-1,3-cyclohexadiene 92.0+%, TCI America™

Catalog No. T17185ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17185ML 5 mL
T17181ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17185ML Supplier TCI America Supplier No. T17185ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4724-89-4
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00045518
InChI Key SZHAWDAGEJWQJK-UHFFFAOYSA-N
PubChem CID 78453
IUPAC Name 1,3,5,5-tetramethylcyclohexa-1,3-diene
SMILES CC1=CC(=CC(C1)(C)C)C

Specifications

Color Yellow
Quantity 5 mL
UN Number 3295
Synonym 1,3,5,5-tetramethyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1,3,5,5-tetramethyl, acmc-1alu2, 1, 1,3,5,5-tetramethyl, 1,3,5,5-tetramethylcyclohexadiene, 1,5,5-tetramethyl-1,3-cyclohexadiene, 1,3-cyclohexadiene,1,3,5,5-tetramethyl
Formula Weight 136.24
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5,5-Tetramethyl-1,3-cyclohexadiene