missing translation for 'onlineSavingsMsg'
Learn More

1-(3,4-Dichlorobenzyl)piperazine, 97%

Catalog No. p-7052758
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5568506 5 g
AAH5568514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5568506 Supplier Thermo Scientific Chemicals Supplier No. H5568506

CAS: 55513-17-2 | C11H14Cl2N2 | 245.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55513-17-2
Molecular Formula C11H14Cl2N2
Molecular Weight (g/mol) 245.147
MDL Number MFCD03407488
InChI Key PNDSYXGJCWKNFG-UHFFFAOYSA-N
Synonym 1-3,4-dichlorobenzyl piperazine, 1-3,4-dichlorophenyl methyl piperazine, piperazine, 1-3,4-dichlorophenyl methyl, 1-3,4-dichloro-benzyl-piperazine, acmc-20apc5, 3,4-dichlorobenzylpiperazine, 1-3,4-dichlorobenyl piperazine, 3,4-dichlorophenyl methyl piperazine
PubChem CID 2758074
IUPAC Name 1-[(3,4-dichlorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC(=C(C=C2)Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.225 g/mL
Boiling Point 124°C to 128°C (0.02 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 245.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(3,4-Dichlorobenzyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only