missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,4,6-Tetrakis(methoxymethyl)glycoluril 98.0+%, TCI America™
$139.81 - $398.90
Chemical Identifiers
| CAS | 17464-88-9 |
|---|---|
| Molecular Formula | C12H22N4O6 |
| Molecular Weight (g/mol) | 318.33 |
| MDL Number | MFCD00402438 |
| InChI Key | XGQJGMGAMHFMAO-UHFFFAOYSA-N |
| PubChem CID | 87125 |
| IUPAC Name | 1,3,4,6-tetrakis(methoxymethyl)-octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione |
| SMILES | COCN1C2C(N(COC)C1=O)N(COC)C(=O)N2COC |
Chemical Identifiers
| 17464-88-9 | |
| 318.33 | |
| XGQJGMGAMHFMAO-UHFFFAOYSA-N | |
| 1,3,4,6-tetrakis(methoxymethyl)-octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione |
| C12H22N4O6 | |
| MFCD00402438 | |
| 87125 | |
| COCN1C2C(N(COC)C1=O)N(COC)C(=O)N2COC |
Specifications
| 17464-88-9 | |
| White | |
| MFCD00402438 | |
| XGQJGMGAMHFMAO-UHFFFAOYSA-N | |
| 1,3,4,6-tetrakis(methoxymethyl)-octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione | |
| 87125 | |
| ≥98.0% (N) | |
| 1,3,4,6-Tetrakis(methoxymethyl)glycoluril |
| 118°C | |
| C12H22N4O6 | |
| 5 g | |
| COCN1C2C(N(COC)C1=O)N(COC)C(=O)N2COC | |
| 318.33 | |
| 318.33 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes