missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride

Catalog No. p-7043300
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6697503 1 g
AAJ6697506 5 g
AAJ6697514 25 g
AAJ6697518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6697503 Supplier Thermo Scientific Chemicals Supplier No. J6697503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10034-20-5 | C14H22ClNO9 | 383.778 g/mol

NSC 82044 is a potential metabolic inhibitor of cellular-membrane glycoconjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NSC 82044 is a potential metabolic inhibitor of cellular-membrane glycoconjugates.

Solubility
Soluble in water at 10mg/ml

Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place.

Chemical Identifiers

CAS 10034-20-5
Molecular Formula C14H22ClNO9
Molecular Weight (g/mol) 383.778
MDL Number MFCD01075204
InChI Key BQLUYAHMYOLHBX-LEXQQUKESA-N
Synonym 1,3,4,6-tetra-o-acetyl-2-amino-2-desoxy-d-glucopyranose hydrochloride, tetra-o-acetyl-glucosaminhydrochlorid, 2r,3s,4r,5r-3,4,6-tris acetyloxy-5-aminooxan-2-yl methyl acetate hydrochloride
PubChem CID 57373443
IUPAC Name [(2R,3S,4R,5R)-3,4,6-triacetyloxy-5-aminooxan-2-yl]methyl acetate;hydrochloride
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)N)OC(=O)C)OC(=O)C.Cl

Specifications

Color Brown to White
Quantity 1 g
Solubility Information Soluble in water at 10mg/ml
Formula Weight 383.78
Physical Form Powder
Chemical Name or Material 1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride
RTECSNumber LZ6574000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only