missing translation for 'onlineSavingsMsg'
Learn More

1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC34039DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC34039DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

852227-97-5
287.21
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 3-piperidinophenylboronic acid, pinacol ester, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 3-piperidin-1-yl benzeneboronic acid, pinacol ester, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
C17H26BNO2
IIUXWPXCCXVCPD-UHFFFAOYSA-N
4961251
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N3CCCCC3

Specifications

1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
95%
C17H26BNO2
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 3-piperidinophenylboronic acid, pinacol ester, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 3-piperidin-1-yl benzeneboronic acid, pinacol ester, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N3CCCCC3
287.21
287.21
852227-97-5
Amber Glass Bottle
5g
IIUXWPXCCXVCPD-UHFFFAOYSA-N
1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
4961251
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents