missing translation for 'onlineSavingsMsg'
Learn More

1,3,3-Trimethylindolino-beta-naphthopyrylospiran 98.0+%, TCI America™

Catalog No. T04235G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04235G 5 g
T04231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04235G Supplier TCI America Supplier No. T04235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1592-43-4
Molecular Formula C23H21NO
Molecular Weight (g/mol) 327.427
MDL Number MFCD00059909
InChI Key DTQKEQFXLWFVCS-UHFFFAOYSA-N
PubChem CID 2728827
IUPAC Name 1',3',3'-trimethylspiro[benzo[f]chromene-3,2'-indole]
SMILES CC1(C2=CC=CC=C2N(C13C=CC4=C(O3)C=CC5=CC=CC=C54)C)C

Specifications

Melting Point 174°C
Color Red-White
Quantity 5 g
Synonym 1,3,3-Trimethylspiro[indoline-2,3′-[3H]naphth[2,1-b]pyran]
Formula Weight 327.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,3-Trimethylindolino-beta-naphthopyrylospiran [Photochromic Compound]