missing translation for 'onlineSavingsMsg'
Learn More

1,3,3-Trimethylindolino-8'-methoxybenzopyrylospiran 98.0+%, TCI America™

Catalog No. p-7136268
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04161G 1 g
T041625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04161G Supplier TCI America Supplier No. T04161G
Only null left

Chemical Identifiers

CAS 13433-31-3
Molecular Formula C20H21NO2
Molecular Weight (g/mol) 307.393
MDL Number MFCD00059897
InChI Key NQVQBIMDDKDYAO-UHFFFAOYSA-N
PubChem CID 2841117
IUPAC Name 8-methoxy-1',3',3'-trimethylspiro[chromene-2,2'-indole]
SMILES CC1(C2=CC=CC=C2N(C13C=CC4=C(O3)C(=CC=C4)OC)C)C

Specifications

Melting Point 124°C
Quantity 1 g
Synonym 8-Methoxy-1′C,3′C,3′C-trimethylspiro[1(2H)-benzopyran-2,2′C-indoline]
Formula Weight 307.39
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,3-Trimethylindolino-8′-methoxybenzopyrylospiran [Photochromic Compound]