Learn More
Pendotech Pendotech Electronic Assembly For 1/2 Inch Single Use Rotary Flow Meter
Catalog No. NC1428383
Encompass
Electronic Assembly for one PVDF rotor with 1/2 inch hose barb (includes one rotor), 1.0-20.0 lpm, clip mount. With individual calibrations.
Catalog No. NC1428383 Supplier Pendotech Supplier No. FM23WVE
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Electronic Assembly for one PVDF rotor with 1/2 inch hose barb (includes one rotor), 1.0-20.0 lpm, clip mount. With individual calibrations.