missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thienyl)ethylamine, tech. 90%

Catalog No. AAH26052ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26052ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH26052ME Supplier Thermo Scientific Chemicals Supplier No. H26052ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6309-16-6 | C6H10NS | 128.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6309-16-6
Molecular Formula C6H10NS
Molecular Weight (g/mol) 128.21
MDL Number MFCD02734311
InChI Key LYJBVRVJQXVVPI-RXMQYKEDSA-O
Synonym 1-thiophen-2-yl ethanamine, 1-2-thienyl ethylamine, 1-thiophen-2-yl-ethylamine, 1-thiophen-2-yl ethan-1-amine, 1-thien-2-ylethanamine, 1-2-thienyl ethanamine, 1-2-thienyl ethanamin, 1-thiophen-2-ylethylamine, 1-thien-2-yl ethylamine, 2-1-aminoethyl thiophene
PubChem CID 238233
IUPAC Name 1-thiophen-2-ylethanamine
SMILES C[C@@H]([NH3+])C1=CC=CS1

Specifications

Refractive Index 1.5475
Quantity 500 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 127.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-(2-Thienyl)ethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 679-243-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only