missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thien-2-ylphenyl)methanamine hydrochloride, ≥95%, Thermo Scientific™

Catalog No. p-3176130
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC35563CB 250mg Amber Glass Bottle
CC35563DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC35563CB Supplier Thermo Scientific Chemicals Supplier No. CC35563CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 863991-95-1
Molecular Formula C11H12ClNS
Molecular Weight (g/mol) 225.73
MDL Number MFCD07772821
InChI Key XLJIHGHEJAGJJC-UHFFFAOYSA-N
Synonym 1-2-thien-2-ylphenyl methanamine hydrochloride, 1-2-thien-2-ylphenyl methylamine hydrochloride, 2-thiophen-2-yl phenyl methanamine hydrochloride, 1-2-thien-2-ylphenyl methanaminehydrochloride, 1-2-thiophen-2-yl phenyl methanamine hydrochloride, c11h11ns.hcl, 2-thiophen-2-yl-benzylamine hydrochloride, 2-2-thienyl phenyl methylamine, chloride, 2-thien-2-yl phenyl methylamine hydrochloride
PubChem CID 18525724
IUPAC Name 1-[2-(thiophen-2-yl)phenyl]methanamine hydrochloride
SMILES Cl.NCC1=CC=CC=C1C1=CC=CS1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 225.74
Percent Purity 95%
Chemical Name or Material 1-(2-Thien-2-ylphenyl)methanamine hydrochloride
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.