missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid, 95%

Catalog No. p-7055434
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64012MD 250 mg
AAH6401203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64012MD Supplier Thermo Scientific Chemicals Supplier No. H64012MD
Only null left

CAS: 1141878-45-6 | C9H12BF3N2O3 | 264.011 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1141878-45-6
Molecular Formula C9H12BF3N2O3
Molecular Weight (g/mol) 264.011
MDL Number MFCD17214248
InChI Key IIRVABFYRULQMI-UHFFFAOYSA-N
Synonym 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 1-tetrahydropyran-2-yl-3-trifluoromethyl pyrazole-5-boronic acid, 1-oxan-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 2-tetrahydropyran-2-yl-5-trifluoromethyl pyrazol-3-yl boronic acid, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronicacid, 2-oxan-2-yl-5-trifluoromethyl pyrazol-3-ylboronic acid, acmc-2099kl, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-ylboronic acid, boronic acid, b-1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl
PubChem CID 53216481
IUPAC Name [2-(oxan-2-yl)-5-(trifluoromethyl)pyrazol-3-yl]boronic acid
SMILES B(C1=CC(=NN1C2CCCCO2)C(F)(F)F)(O)O

Specifications

Quantity 250 mg
Formula Weight 264.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only