missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester, 95%

Catalog No. AAH32369ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32369ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH32369ME Supplier Thermo Scientific Chemicals Supplier No. H32369ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1029684-37-4 | C14H23BN2O3 | 278.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1029684-37-4
Molecular Formula C14H23BN2O3
Molecular Weight (g/mol) 278.159
MDL Number MFCD11100960
InChI Key BVIWGBKKGPHNAB-UHFFFAOYSA-N
Synonym 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-imidazole, 1-tetrahydro-2h-pyran-2-yl-1h-imidazole-5-boronic acid pinacol ester, 1-2-tetrahydropyranyl-1h-imidazole-5-boronic acid pinacol ester, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-imidazole, 1-tetrahydro-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-imidazole, 1-tetrahydropyran-2-yl-1h-imidazole-5-boronic ac, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl imidazole, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl imidazole, 1-tetrahydro-pyran-2-yl-1h-imidazole-5-boronic acid pinacol ester, 1-tetrahydropyran-2-yl-1h-imidazole-5-boronic acid pinacol ester
PubChem CID 24880008
IUPAC Name 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)imidazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=CN2C3CCCCO3

Specifications

Melting Point 129°C to 133°C
Quantity 500 mg
Solubility Information Slightly soluble in water.
Formula Weight 278.16
Percent Purity 95%
Chemical Name or Material 1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only