missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Pyrimidyl)piperazine 98.0+%, TCI America™
$44.73 - $128.62
Chemical Identifiers
| CAS | 20980-22-7 |
|---|---|
| Molecular Formula | C8H12N4 |
| Molecular Weight (g/mol) | 164.212 |
| MDL Number | MFCD00040742 |
| InChI Key | MRBFGEHILMYPTF-UHFFFAOYSA-N |
| Synonym | 2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine |
| PubChem CID | 88747 |
| IUPAC Name | 2-piperazin-1-ylpyrimidine |
| SMILES | C1CN(CCN1)C2=NC=CC=N2 |
Chemical Identifiers
| 20980-22-7 | |
| 164.212 | |
| MRBFGEHILMYPTF-UHFFFAOYSA-N | |
| 88747 | |
| C1CN(CCN1)C2=NC=CC=N2 |
| C8H12N4 | |
| MFCD00040742 | |
| 2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine | |
| 2-piperazin-1-ylpyrimidine |
Specifications
| 20980-22-7 | |
| White-Yellow | |
| C8H12N4 | |
| 5 g | |
| MRBFGEHILMYPTF-UHFFFAOYSA-N | |
| 2-piperazin-1-ylpyrimidine | |
| 88747 | |
| ≥98.0% (GC) | |
| 1-(2-Pyrimidyl)piperazine |
| 33°C | |
| 120°C | |
| MFCD00040742 | |
| 2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine | |
| C1CN(CCN1)C2=NC=CC=N2 | |
| 164.212 | |
| 164.21 | |
| Crystalline Lumps |
Safety and Handling
RTECSNumber : UV9702000
TSCA : No