missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridylazo)-2-naphthol, 98%

Catalog No. AAA1747503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1747503 1 g
AAA1747506 5 g
AAA1747514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1747503 Supplier Thermo Scientific Chemicals Supplier No. A1747503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85-85-8 | C15H11N3O | 249.273 g/mol

1-(2-Pyridylazo)-2-naphthol is widely used as an indicator for complexometry. It is used as a reagent for the extraction and spectrophotometric determination (for colorimetric analysis) of many transition metal elements like arsenic, chromium, cobalt, copper, cadmium, manganese, zinc and zirconium. It is also used for the analysis of uranium and thorium in solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Pyridylazo)-2-naphthol is widely used as an indicator for complexometry. It is used as a reagent for the extraction and spectrophotometric determination (for colorimetric analysis) of many transition metal elements like arsenic, chromium, cobalt, copper, cadmium, manganese, zinc and zirconium. It is also used for the analysis of uranium and thorium in solutions.

Solubility
Insoluble in water.

Notes
Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 85-85-8
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.273
MDL Number MFCD00004071
InChI Key RAXUMGMWXZYADR-OBGWFSINSA-N
Synonym 1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl
PubChem CID 5376264
IUPAC Name (1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3

Specifications

Melting Point 138°C to 141°C
Quantity 1 g
Beilstein 206390
Solubility Information Insoluble in water.
Formula Weight 249.27
Percent Purity 98%
Chemical Name or Material 1-(2-Pyridylazo)-2-naphthol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-637-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only