missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridyl)piperazine 98.0+%, TCI America™

Catalog No. p-7134913
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
P10335G 5 g
P103325G 25 g
2 options

Catalog No. P10335G

Supplier: TCI America P10335G

Only null left

Chemical Identifiers

CAS 34803-66-2
Molecular Formula C9H13N3
Molecular Weight (g/mol) 163.224
MDL Number MFCD00006216
InChI Key GZRKXKUVVPSREJ-UHFFFAOYSA-N
Synonym 1-2-pyridyl piperazine, 1-pyridin-2-yl piperazine, 1-pyridin-2-yl-piperazine, 1-2-pyridinyl piperazine, piperazine, 1-2-pyridinyl, 2-pyridylpiperazine, pyridinylpiperazine, 2-1-piperazinyl pyridine
PubChem CID 94459
IUPAC Name 1-pyridin-2-ylpiperazine
SMILES C1CN(CCN1)C2=CC=CC=N2

Specifications

Boiling Point 122°C
Quantity 5 g
Formula Weight 163.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Pyridyl)piperazine