missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridyl)piperazine, 99%

Catalog No. AAL0545106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0545106 5 g
AAL0545114 25 g
AAL0545122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0545106 Supplier Thermo Scientific Chemicals Supplier No. L0545106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34803-66-2 | C9H13N3 | 163.224 g/mol

1-(2-Pyridyl)piperazine may be employed as reagent for the determination of both aliphatic and aromatic isocyanates in air by reversed-phase HPLC. It may be employed as reagent for the fluorometric determination of airborne diisocyantes. It is a compound that belongs to a class of selective α2-adrenoceptor antagonists. 1-(2-Pyridyl)piperazine shows sympatholytic activity. 1-(2-Pyridyl)piperazine is also a metabolite of Azaperone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Pyridyl)piperazine may be employed as reagent for the determination of both aliphatic and aromatic isocyanates in air by reversed-phase HPLC. It may be employed as reagent for the fluorometric determination of airborne diisocyantes. It is a compound that belongs to a class of selective α2-adrenoceptor antagonists. 1-(2-Pyridyl)piperazine shows sympatholytic activity. 1-(2-Pyridyl)piperazine is also a metabolite of Azaperone.

Solubility
Soluble in Chloroform, Methanol.

Notes
Air Sensitive. Store away from air and oxidizing agents. Incompatible with carbondioxide, Strong acids, Acid chlorides, Acid anhydrides.

Chemical Identifiers

CAS 34803-66-2
Molecular Formula C9H13N3
Molecular Weight (g/mol) 163.224
MDL Number MFCD00006216
InChI Key GZRKXKUVVPSREJ-UHFFFAOYSA-N
Synonym 1-2-pyridyl piperazine, 1-pyridin-2-yl piperazine, 1-pyridin-2-yl-piperazine, 1-2-pyridinyl piperazine, piperazine, 1-2-pyridinyl, 2-pyridylpiperazine, pyridinylpiperazine, 2-1-piperazinyl pyridine
PubChem CID 94459
IUPAC Name 1-pyridin-2-ylpiperazine
SMILES C1CN(CCN1)C2=CC=CC=N2

Specifications

Density 1.072
Boiling Point 120°C to 122°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 5 g
UN Number UN2735
Beilstein 140423
Sensitivity Air Sensitive
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 163.22
Percent Purity 99%
Chemical Name or Material 1-(2-Pyridyl)piperazine
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 252-220-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only