missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyrazinyl)piperazine, 98%

Catalog No. p-2689369
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC367130010 1 g Glass bottle
AC367130050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC367130010 Supplier Thermo Scientific Chemicals Supplier No. 367130010
Only null left

CAS: 34803-68-4 | C8H12N4 | 164.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34803-68-4
Molecular Formula C8H12N4
Molecular Weight (g/mol) 164.21
MDL Number MFCD00040766
InChI Key HCGFLVDMFDHYJD-UHFFFAOYSA-N
Synonym 2-piperazin-1-yl pyrazine, 1-2-pyrazinyl-piperazine, 1-piperazinylpyrazine, 1-pyrazin-2-yl piperazine, 1-2-pyrazinyl piperazine, pyrazin-2-yl-piperazine, 3,4,5,6-tetrahydro-2h-1,2' bipyrazinyl, 2-piperazinylpyrazine, 2-piperazinopyrazine, 1-piperazinyl pyrazine
PubChem CID 2734639
IUPAC Name 2-piperazin-1-ylpyrazine
SMILES C1CN(CCN1)C2=NC=CN=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Boiling Point 105°C to 108°C (0.5 Mbar)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 164.21
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-(2-Pyrazinyl)piperazine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 252-221-9

RUO – Research Use Only