missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanedithiol 95.0+%, TCI America™

Catalog No. p-7134790
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08215ML 5 mL
P082125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08215ML Supplier TCI America Supplier No. P08215ML
Only null left

Chemical Identifiers

CAS 814-67-5
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.22
MDL Number MFCD00022073
InChI Key YGKHJWTVMIMEPQ-UHFFFAOYNA-N
Synonym 1,2-Dimercaptopropane, Propylenedithiol
PubChem CID 61217
IUPAC Name propane-1,2-dithiol
SMILES CC(S)CS

Specifications

Color Yellow
Quantity 5 mL
UN Number 1228
Formula Weight 108.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Propanedithiol
RTECSNumber TZ2585000
TSCA Yes