missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol 99.0+%, TCI America™

Catalog No. P0485500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0485500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0485500G Supplier TCI America Supplier No. P0485500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00064272
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Melting Point -59°C
Color Colorless
Boiling Point 188°C
Quantity 500 g
Formula Weight 76.10
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Propanediol
EINECSNumber (2)-0234
RTECSNumber TY2000000
TSCA Yes