missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol, Reagent, ACS, 99.5%, Spectrum™ Chemical

P1440, 57-55-6, C3H8O2

Supplier:  Spectrum Chemical Mfg Cor P1440500MLGL

Quantity 500 mL
Packaging Amber Glass Bottle

Catalog No. 18600039


Get Up to $500 in Chemicals with Purchase
View all promotions

Description

Description

Spectrum™ Chemical 1,2-Propanediol, Reagent, ACS or propylene glycol is an organic compound and colorless liquid without odor. It has a slightly sweet taste and chemically classed as a diol. It is considered generally recognized as safe (GRAS) by the U.S. Food and Drug Administration, and it is used as an humectant (E1520), solvent, and preservative in food and for tobacco products, as well as being one of the major ingredients of the "e-liquid" used in electronic cigarettes along with vegetable glycerin. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications

Specifications

57-55-6
Amber Glass Bottle
500 mL
CC(O)CO
76.10
ACS
100%
C3H8O2
DNIAPMSPPWPWGF-UHFFFAOYNA-N
propane-1,2-diol
99.5%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions