missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol monomethyl ether acetate, 99%, stab. with 50ppm BHT

CAS: 108-65-6 | C6H12O3 | 132.159 g/mol

Supplier:  Thermo Scientific Chemicals L15459AU

 View more versions of this product

Catalog No. AAL15459AU

Only null left
Explore available promotions

Description

Description

1,2-Propanediol monomethyl ether acetate is used as a solvent for paints, inks, lacquers, varnishes, cleaners and coatings. It is also used as a solvent for degreasing circuit boards and in food contact applications. Further, it is used in photoresist formulations in the semiconductor industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Propanediol monomethyl ether acetate is used as a solvent for paints, inks, lacquers, varnishes, cleaners and coatings. It is also used as a solvent for degreasing circuit boards and in food contact applications. Further, it is used in photoresist formulations in the semiconductor industry.

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, reducing agents, acids and alkalis.
Specifications

Specifications

1,2-Propanediol monomethyl ether acetate
108-65-6
0.968
145°C to 146°C
Ester-like
1.402
1000 mL
1751656
Miscible with water.
CC(COC)OC(=O)C
132.159
132.16
stabilized with 50ppm BHT
-67°C
4
43°C (109°F)
C6H12O3
MFCD00038500
UN3272
1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester
LLHKCFNBLRBOGN-UHFFFAOYSA-N
1-methoxypropan-2-yl acetate
7946
99%

Chemical Identifiers

108-65-6
132.159
LLHKCFNBLRBOGN-UHFFFAOYSA-N
7946
CC(COC)OC(=O)C
C6H12O3
MFCD00038500
1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester
1-methoxypropan-2-yl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 203-603-9

RTECSNumber : AI8920000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only