missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol monomethyl ether acetate, 99%, stab. with 50ppm BHT

Catalog No. AAL15459AE
Change view
Click to view available options
Quantity:
100 mL
1000 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15459AE 100 mL
AAL15459AU 1000 mL
AAL154590F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL15459AE Supplier Thermo Scientific Chemicals Supplier No. L15459AE

CAS: 108-65-6 | C6H12O3 | 132.159 g/mol

1,2-Propanediol monomethyl ether acetate is used as a solvent for paints, inks, lacquers, varnishes, cleaners and coatings. It is also used as a solvent for degreasing circuit boards and in food contact applications. Further, it is used in photoresist formulations in the semiconductor industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Propanediol monomethyl ether acetate is used as a solvent for paints, inks, lacquers, varnishes, cleaners and coatings. It is also used as a solvent for degreasing circuit boards and in food contact applications. Further, it is used in photoresist formulations in the semiconductor industry.

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, reducing agents, acids and alkalis.

Chemical Identifiers

CAS 108-65-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00038500
InChI Key LLHKCFNBLRBOGN-UHFFFAOYSA-N
Synonym 1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester
PubChem CID 7946
IUPAC Name 1-methoxypropan-2-yl acetate
SMILES CC(COC)OC(=O)C

Specifications

Melting Point -67°C
Density 0.968
pH 4
Boiling Point 145°C to 146°C
Flash Point 43°C (109°F)
Odor Ester-like
Refractive Index 1.402
Quantity 100 mL
UN Number UN3272
Beilstein 1751656
Solubility Information Miscible with water.
Formula Weight 132.16
Percent Purity 99%
Chemical Name or Material 1,2-Propanediol monomethyl ether acetate, stabilized with 50ppm BHT
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 203-603-9
RTECSNumber AI8920000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only