missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Propanediol, MilliporeSigma™
Specifications
Specifications
| Melting Point | -60°C |
| Boiling Point | 187.4°C |
| Flash Point | 98.85°C (209.9°F) |
| Packaging | Poly Pail |
| Quantity | 18.9 L |
| Solubility Information | Water: Soluble |
| Formula Weight | 76.1 |
| Percent Purity | ≥98.0% |
| Grade | ACS |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Recommended Storage | Room Temperature. Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage. |