missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol, MilliporeSigma™

Catalog No. MPX16903
Change view
Click to view available options
Quantity:
4 L
18.9 L
Packaging:
Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX16903 4 L Glass Bottle
MPX16905 18.9 L Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MPX16903 Supplier Technidata Supplier No. PX16903

For General Chemistry

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Melting Point -60°C
Boiling Point 187.4°C
Flash Point 98.85°C (209.9°F)
Packaging Glass Bottle
Quantity 4 L
Solubility Information Water: Soluble
Formula Weight 76.1
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 1,2-Propanediol
Recommended Storage Room Temperature. Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.