missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol, 98+%

Catalog No. p-7024157
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1840636 500 g
AAA184060E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1840636 Supplier Thermo Scientific Chemicals Supplier No. A1840636
Only null left

CAS: 57-55-6 | C3H8O2 | 76.095 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00064272
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Melting Point -60°C
Density 1.036
pH Neutral
Boiling Point 186°C to 188°C
Flash Point 107°C (225°F)
Odor Nearly Odorless
Refractive Index 1.432
Quantity 500 g
Beilstein 1340498
Sensitivity Hygroscopic
Merck Index 14,7855
Solubility Information Miscible with water,benzene,acetone,chloroform and ether. Immiscible with fixed oils.
Formula Weight 76.1
Percent Purity ≥98%
Chemical Name or Material 1,2-Propanediol
Show More Show Less
EINECSNumber 200-338-0
RTECSNumber TY2000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only