missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich 1,2-Propanediol
Description
1,2-Propanediol
Specifications
Specifications
| CAS | 57-55-6 |
| Melting Point | -60°C |
| Density | 1.036 g/mL at 25°C |
| Boiling Point | 187°C |
| Molecular Formula | C3H8O2 |
| Refractive Index | n20/D 1.432 |
| Linear Formula | CH3CH(OH)CH2OH |
| MDL Number | MFCD00064272 |
| Quantity | 2 L |
| Synonym | 1,2-PDO; Propylene glycol |
| Show More |