missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediacetic Acid 98.0+%, TCI America™

Catalog No. P10525G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10525G Supplier TCI America Supplier No. P10525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7500-53-0
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004329
InChI Key MMEDJBFVJUFIDD-UHFFFAOYSA-N
Synonym 1,2-phenylenediacetic acid, 2,2'-1,2-phenylene diacetic acid, o-phenylenediacetic acid, o-benzenediacetic acid, 1,2-benzenediacetic acid, 2-2-carboxymethyl phenyl acetic acid, phenylenediacetic acid, acmc-209owx, 1,2-benzenediaceticacid, o-phenylene-diacetic acid
PubChem CID 24123
IUPAC Name 2-[2-(carboxymethyl)phenyl]acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)CC(=O)O

Specifications

Melting Point 150°C
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Phenylenediacetic Acid