missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediacetic acid, 98+%

Catalog No. AAA1385106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1385106 5 g
AAA1385114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1385106 Supplier Thermo Scientific Chemicals Supplier No. A1385106

CAS: 7500-53-0 | C10H10O4 | 194.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7500-53-0
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004329
InChI Key MMEDJBFVJUFIDD-UHFFFAOYSA-N
Synonym 1,2-phenylenediacetic acid, 2,2'-1,2-phenylene diacetic acid, o-phenylenediacetic acid, o-benzenediacetic acid, 1,2-benzenediacetic acid, 2-2-carboxymethyl phenyl acetic acid, phenylenediacetic acid, acmc-209owx, 1,2-benzenediaceticacid, o-phenylene-diacetic acid
PubChem CID 24123
IUPAC Name 2-[2-(carboxymethyl)phenyl]acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)CC(=O)O

Specifications

Melting Point 150°C to 152°C
Odor Odorless
Quantity 5 g
Beilstein 2616747
Formula Weight 194.19
Percent Purity ≥98%
Chemical Name or Material 1,2-Phenylenediacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-355-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only