missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Phenyl-1,3-thiazol-4-yl)ethan-1-one, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 95.0 |
| Assay Percent Range | 95% |
| Quantity | 10g |
| Formula Weight | 203.26 |
| Percent Purity | 95% |
| Chemical Name or Material | 1-(2-Phenyl-1, 3-thiazol-4-yl)ethan-1-one |