missing translation for 'onlineSavingsMsg'
Learn More

1,2-Octanediol 96.0+%, TCI America™

Catalog No. p-7134271
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O027725G 25 g
O0277400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O027725G Supplier TCI America Supplier No. O027725G
Only null left

Chemical Identifiers

CAS 1117-86-8
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00010738
InChI Key AEIJTFQOBWATKX-UHFFFAOYSA-N
Synonym 1,2-octanediol, 1,2-dihydroxyoctane, 1,2-octylene glycol, capyryl glycol, racemic 1,2-octanediol, acmc-2099as, dsstox_cid_16646, dsstox_rid_79289
PubChem CID 14231
ChEBI CHEBI:34056
IUPAC Name octane-1,2-diol
SMILES CCCCCCC(CO)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 146.23
Percent Purity ≥96.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,2-Octanediol
EINECSNumber (2)-0240
TSCA Yes