Learn More
1,2-Octanediol, 97%
Description
1,2-Octanediol was used as organic modifier to improve the HPLC separation of organic acids and bases. It was also used in preparation of halohydrin palmitates. It is useful for treating head louse infestation clinically. It is also used in coating materials, slurries, paper mills and water circulation systems for the effective preservation against bacteria and fungi. It is used as an emollient, humectant, and wetting agent in cosmetic and skin care products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water (3 g/L) at 20°C.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 28°C to 34°C |
| Boiling Point | 131°C to 133°C (10 mmHg) |
| Flash Point | 138°C (280°F) |
| Quantity | 50 g |
| Beilstein | 1719619 |
| Solubility Information | Soluble in water (3g/L) at 20°C. |
| Formula Weight | 146.23 |
| Percent Purity | 97% |
| Chemical Name or Material | 1,2-Octanediol |
Safety and Handling
| Hazard Category | H319 |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 214-254-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only