missing translation for 'onlineSavingsMsg'
Learn More

1,2-O-Isopropylidene-alpha-D-xylofuranose 98.0+%, TCI America™

Catalog No. p-7131560
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07215G 5 g
I072125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07215G Supplier TCI America Supplier No. I07215G
Only null left

Chemical Identifiers

CAS 20031-21-4
Molecular Formula C8H14O5
Molecular Weight (g/mol) 190.195
MDL Number MFCD00063295
InChI Key JAUQZVBVVJJRKM-XZBKPIIZSA-N
Synonym 1,2-o-isopropylidene-alpha-d-xylofuranose, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2-o-isopropylidene-a-d-xylofuranose, alpha-d-xylofuranose, 1,2-o-1-methylethylidene, 1,2-o-isopropylidene-d-xylofuranose, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 2,3-d 1,3 dioxol-6-ol, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, .alpha.-d-xylofuranose, 1,2-o-1-methylethylidene, 1,2-o-isopropylidene-, a-d-xylofuranose
PubChem CID 88338
IUPAC Name (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OC2C(C(OC2O1)CO)O)C

Specifications

Melting Point 70°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,2-O-Isopropylidene-alpha-D-xylofuranose