missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-O-Isopropylidene-alpha-D-xylofuranose 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 70°C |
| Color | White |
| Quantity | 25 g |
| Physical Form | Crystal-Powder at 20°C |
| Formula Weight | 190.20 |
| Percent Purity | ≥98.0% (GC) |
| Chemical Name or Material | 1,2-O-Isopropylidene-alpha-D-xylofuranose |
Safety and Handling
| TSCA | Yes |