missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2-O-Isopropylidene-alpha-D-xylofuranose, 99%

Catalog No. AC348881000
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC348881000 100 g Glass bottle
AC348880100 10 g Glass bottle
AC348880250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC348881000 Supplier Thermo Scientific Chemicals Supplier No. 348881000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20031-21-4 | C8H14O5 | 190.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20031-21-4
Molecular Formula C8H14O5
Molecular Weight (g/mol) 190.2
MDL Number MFCD00063295
InChI Key JAUQZVBVVJJRKM-XZBKPIIZSA-N
Synonym 1,2-o-isopropylidene-alpha-d-xylofuranose, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2-o-isopropylidene-a-d-xylofuranose, alpha-d-xylofuranose, 1,2-o-1-methylethylidene, 1,2-o-isopropylidene-d-xylofuranose, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 2,3-d 1,3 dioxol-6-ol, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 3ar,5r,6s,6ar-5-hydroxymethyl-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, .alpha.-d-xylofuranose, 1,2-o-1-methylethylidene, 1,2-o-isopropylidene-, a-d-xylofuranose
PubChem CID 88338
IUPAC Name (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OC2C(C(OC2O1)CO)O)C

Specifications

Melting Point 68.0°C to 70.0°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Quantity 100 g
Boiling Point 112.0°C to 114.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Waxy Solid or Coarse Crystals and Chunks
Assay Percent Range 98.5% min. (GC)
Beilstein 19, IV, 4834
Specific Rotation Condition − 18.00 (20.00°C c=1,water)
Specific Rotation − 18.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,dmso,dmf,chcl3,,acetone and ether
Formula Weight 190.2
Percent Purity 99%
Chemical Name or Material 1, 2-O-Isopropylidene-α-D-xylofuranose, 99%
Show More Show Less
Hazard Category HYGROSCOPIC
TSCA TSCA

RUO – Research Use Only