missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(Morpholin-4-yl)ethyl]piperazine, 99%

Catalog No. p-2689499
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC367140010 1 g Glass bottle
AC367140050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC367140010 Supplier Thermo Scientific Chemicals Supplier No. 367140010

CAS: 4892-89-1 | C10H21N3O | 199.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4892-89-1
Molecular Formula C10H21N3O
Molecular Weight (g/mol) 199.29
MDL Number MFCD00191217
InChI Key SAJZEJMFAWZNCQ-UHFFFAOYSA-N
Synonym 4-2-piperazin-1-yl-ethyl-morpholine, 4-2-piperazin-1-yl ethyl morpholine, 4-2-piperazin-1-ylethyl morpholine, 1-2-morpholin-4-yl-ethyl-piperazine, 1-2-morpholinoethyl-piperazine, 1-2-morpholin-4-yl ethyl piperazine, morpholine, 4-2-1-piperazinyl ethyl, 1-2-morpholinoethyl piperazine, 4-2-1-piperazinyl ethyl morpholine
PubChem CID 2734640
IUPAC Name 4-(2-piperazin-1-ylethyl)morpholine
SMILES C1CN(CCN1)CCN2CCOCC2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White to Yellow
Boiling Point 82°C to 84°C (0.5 mbar)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 199.29
Percent Purity 99%
Physical Form Oil or Low Melting Crystals
Chemical Name or Material 1-[2-(Morpholin-4-yl)ethyl]piperazine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only