missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Methylenedioxybenzene 99.0+%, TCI America™
Supplier: TCI America M0658100ML
Specifications
| 1,2-Methylenedioxybenzene | |
| -10°C | |
| 172°C | |
| MFCD00005818 | |
| 1993 | |
| FTNJQNQLEGKTGD-UHFFFAOYSA-N | |
| 1,3-benzodioxole | |
| 9229 | |
| 122.12 | |
| Liquid |
| 274-09-9 | |
| Yellow | |
| C7H6O2 | |
| 100 mL | |
| 1,2-methylenedioxybenzene, benzodioxole, benzo d 1,3 dioxole, 1,3-dioxaindan, 3,4-methylenedioxybenzene, 2h-1,3-benzodioxole, 1,2-methylenedioxy benzene, 1,3-dioxindan, methylenedioxybenzene, o-methylenedioxy benzene | |
| C1OC2=CC=CC=C2O1 | |
| 122.123 | |
| CHEBI:38732 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 274-09-9 | |
| 122.123 | |
| FTNJQNQLEGKTGD-UHFFFAOYSA-N | |
| 9229 | |
| 1,3-benzodioxole |
| C7H6O2 | |
| MFCD00005818 | |
| 1,2-methylenedioxybenzene, benzodioxole, benzo d 1,3 dioxole, 1,3-dioxaindan, 3,4-methylenedioxybenzene, 2h-1,3-benzodioxole, 1,2-methylenedioxy benzene, 1,3-dioxindan, methylenedioxybenzene, o-methylenedioxy benzene | |
| CHEBI:38732 | |
| C1OC2=CC=CC=C2O1 |
Safety and Handling
EINECSNumber : (5)-5342
RTECSNumber : DA5600000
TSCA : Yes