missing translation for 'onlineSavingsMsg'
Learn More

1,2-Methylenedioxybenzene 99.0+%, TCI America™

Catalog No. M0658500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0658500ML 500 mL
M065825ML 25 mL
M0658100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0658500ML Supplier TCI America Supplier No. M0658500ML

Chemical Identifiers

CAS 274-09-9
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00005818
InChI Key FTNJQNQLEGKTGD-UHFFFAOYSA-N
Synonym 1,2-methylenedioxybenzene, benzodioxole, benzo d 1,3 dioxole, 1,3-dioxaindan, 3,4-methylenedioxybenzene, 2h-1,3-benzodioxole, 1,2-methylenedioxy benzene, 1,3-dioxindan, methylenedioxybenzene, o-methylenedioxy benzene
PubChem CID 9229
ChEBI CHEBI:38732
IUPAC Name 1,3-benzodioxole
SMILES C1OC2=CC=CC=C2O1

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 172°C
Quantity 500 mL
UN Number 1993
Formula Weight 122.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Methylenedioxybenzene
EINECSNumber (5)-5342
RTECSNumber DA5600000
TSCA Yes